Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGLYRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052890.3 | NP_443122.3 | 2 | 403 | ||
XM_006722633.3 | XP_006722696.1 | 2 | 400 | |||
GenBank mRNA | AB073610.1 | - | 2 | 352 | ||
AF384856.1 | - | 2 | 296 | |||
AK055882.1 | - | 2 | 400 | |||
AK289415.1 | - | 2 | 336 | |||
AK292292.1 | - | 2 | 751 | |||
AY358156.1 | - | 2 | 327 | |||
BC136551.1 | - | 2 | 292 | 9020541 | ||
BC136552.1 | - | 2 | 292 | 9020542 | ||
BC144238.1 | - | 2 | 292 | 9052758 | ||
DB041324.1 | - | 2 | 403 |