Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013653.2 | NP_001013675.1 | 2 | 1170 | ||
GenBank mRNA | CB160046.1 | - | 2 | 550 |