Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C21orf91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100420.1 | NP_001093890.1 | 2 | 200 | ||
NM_001100421.1 | NP_001093891.1 | 2 | 200 | |||
NM_017447.3 | NP_059143.3 | 2 | 200 | |||
GenBank mRNA | AF239726.1 | - | 2 | 153 | ||
AK023825.1 | - | 2 | 165 | |||
AK225123.1 | - | 2 | 171 | |||
AK314471.1 | - | 2 | 173 | |||
BC015468.2 | - | 1 | 116 | 3916462 | ||
BF739854.1 | - | 2 | 199 | |||
CR457317.1 | - | 1 | 109 | |||
CR749496.1 | - | 2 | 180 | |||
DA215855.1 | - | 2 | 173 |