Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313896.1 | NP_001300825.1 | 17 | 2210 | ||
NM_001313982.1 | NP_001300911.1 | 17 | 2198 | |||
NM_024011.3 | NP_076916.2 | 17 | 2201 | |||
NM_033529.3 | NP_277071.2 | 17 | 2171 | |||
NR_132739.1 | 17 | 2085 | ||||
NR_132740.1 | 17 | 2124 | ||||
GenBank mRNA | AF067518.1 | - | 17 | 1834 | ||
AF067519.1 | - | 17 | 1950 | |||
AF067520.1 | - | 17 | 1947 | |||
AF067521.1 | - | 17 | 1920 | |||
AF067522.1 | - | 17 | 1959 | |||
AF067523.1 | - | 17 | 1873 | |||
BC014464.1 | - | 9 | 932 | |||
BC094827.1 | - | 9 | 965 | |||
BC110905.1 | - | 12 | 1162 | 6151813 | ||
U04819.1 | - | 10 | 1274 | |||
U07704.1 | - | 17 | 1982 | |||
U07705.1 | - | 17 | 1976 |