Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSIG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100431.1 | NP_001093901.1 | 5 | 776 | ||
NM_001184830.1 | NP_001171759.1 | 6 | 1058 | |||
NM_001184831.1 | NP_001171760.1 | 5 | 776 | |||
NM_001257403.1 | NP_001244332.1 | 6 | 1058 | |||
NM_007268.2 | NP_009199.1 | 6 | 1058 | |||
XM_017029251.1 | XP_016884740.1 | 5 | 735 | |||
GenBank mRNA | AJ132502.1 | - | 6 | 976 | ||
AK075153.1 | - | 6 | 1013 | |||
AK075171.1 | - | 6 | 1040 | |||
AK293215.1 | - | 6 | 952 | |||
AK297050.1 | - | 6 | 1013 | |||
AK298389.1 | - | 5 | 729 | |||
AK301417.1 | - | 5 | 731 | |||
AL542949.3 | - | 5 | 710 | |||
AL552873.3 | - | 5 | 720 | |||
AY358341.1 | - | 6 | 1049 | |||
BC010525.1 | - | 6 | 934 | 4151604 |