Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTFR1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099625.1 | NP_001093095.1 | 5 | 517 | ||
NM_001099626.1 | NP_001093096.1 | 5 | 569 | |||
NM_001099627.1 | NP_001093097.1 | 5 | 517 | |||
NM_019557.5 | NP_062457.3 | 5 | 615 | |||
GenBank mRNA | AB075880.1 | - | 5 | 445 | ||
AF173891.1 | - | 5 | 418 | |||
AK056721.1 | - | 4 | 1880 | |||
AK127164.1 | - | 1 | 1051 | |||
AK290744.1 | - | 4 | 313 | |||
AK299221.1 | - | 5 | 460 | |||
AK314297.1 | - | 5 | 445 | |||
AL512766.1 | - | 5 | 617 | |||
AY007100.1 | - | 5 | 495 | |||
BC017175.2 | - | 5 | 569 | 3162883 | ||
BI545005.1 | - | 5 | 499 | |||
BI769886.1 | - | 5 | 580 | |||
BX647921.1 | - | 5 | 474 | |||
DB170641.1 | - | 5 | 517 | |||
KU178723.1 | - | 4 | 307 | |||
KU178724.1 | - | 4 | 330 |