Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNCX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080461.1 | NP_001073930.1 | 3 | 491 | ||
GenBank mRNA | KM587717.1 | - | 3 | 1127 |