Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CISD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136498.1 | NP_001129970.1 | 3 | 276 | ||
GenBank mRNA | AK097047.1 | - | 2 | 527 | ||
AK098433.1 | - | 1 | 57 |