Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC129 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257967.1 | NP_001244896.1 | 13 | 2787 | ||
NM_001257968.1 | NP_001244897.1 | 12 | 2812 | |||
NM_194300.3 | NP_919276.2 | 11 | 2675 | |||
NR_047565.1 | 12 | 3189 | ||||
XM_011515213.1 | XP_011513515.1 | 12 | 2779 | |||
XM_017011871.1 | XP_016867360.1 | 12 | 2772 | |||
XM_017011872.1 | XP_016867361.1 | 12 | 2779 | |||
GenBank mRNA | AK095663.1 | - | 10 | 2548 | ||
AK128026.1 | - | 12 | 3189 | |||
AK295011.1 | - | 12 | 2730 | |||
AK304387.1 | - | 13 | 2787 | |||
BC132719.1 | - | 12 | 2678 | 40146741 |