Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002160.3 | NP_002151.2 | 17 | 5479 | ||
XM_005251972.3 | XP_005252029.1 | 16 | 5156 | |||
XM_005251974.3 | XP_005252031.1 | 11 | 3791 | |||
XM_006717096.3 | XP_006717159.1 | 18 | 5705 | |||
XM_006717097.3 | XP_006717160.1 | 16 | 5156 | |||
XM_006717098.3 | XP_006717161.1 | 15 | 4883 | |||
XM_011518626.2 | XP_011516928.1 | 14 | 4613 | |||
XM_011518629.2 | XP_011516931.1 | 12 | 4064 | |||
XM_017014678.1 | XP_016870167.1 | 19 | 5978 | |||
XM_017014679.1 | XP_016870168.1 | 18 | 5705 | |||
XM_017014680.1 | XP_016870169.1 | 18 | 5702 | |||
XM_017014681.1 | XP_016870170.1 | 15 | 4886 | |||
GenBank mRNA | AB210038.1 | - | 17 | 5355 | ||
GU473235.1 | - | 12 | 3978 | |||
M24630.1 | - | 15 | 3777 | |||
M55618.1 | - | 16 | 5202 | |||
X56160.1 | - | 16 | 5118 | |||
X78565.1 | - | 17 | 5380 |