Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271427.1 | NP_001258356.1 | 11 | 1168 | ||
NM_001271428.1 | NP_001258357.1 | 11 | 1171 | |||
NM_001271429.1 | NP_001258358.1 | 10 | 1105 | |||
NM_015047.2 | NP_055862.1 | 11 | 1171 | |||
XM_005245787.1 | XP_005245844.1 | 11 | 1166 | |||
XM_005245788.1 | XP_005245845.1 | 11 | 1163 | |||
GenBank mRNA | AK075563.1 | - | 10 | 1068 | ||
AK291618.1 | - | 11 | 1134 | |||
BC034589.1 | - | 11 | 1164 | 4831005 | ||
BX648627.1 | - | 11 | 1150 | |||
BX648708.1 | - | 11 | 1129 | |||
D42044.2 | - | 11 | 1115 |