Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIFO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300831.1 | NP_001287760.1 | 3 | 355 | ||
NM_181643.5 | NP_857594.2 | 4 | 454 | |||
XM_005270472.1 | XP_005270529.1 | 4 | 563 | |||
XM_017000322.1 | XP_016855811.1 | 3 | 439 | |||
GenBank mRNA | AK074433.1 | - | 4 | 389 | ||
AK303897.1 | - | 3 | 316 | |||
BC029237.1 | - | 3 | 343 | |||
BC068603.1 | - | 4 | 378 | |||
BC093891.1 | - | 4 | 441 | 7939736 | ||
BC101501.1 | - | 4 | 442 | 8069007 | ||
BF674731.1 | - | 4 | 460 | |||
HM237139.1 | - | 4 | 441 | |||
HM237140.1 | - | 4 | 367 |