Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020919.3 | NP_065970.2 | 23, 24 | 4051 | ||
XM_006712654.2 | XP_006712717.1 | 23, 24 | 4628 | |||
XM_006712655.3 | XP_006712718.1 | 19, 20 | 2265 | |||
XM_011511530.2 | XP_011509832.1 | 22, 23 | 3763 | |||
XM_017004569.1 | XP_016860058.1 | 22, 23 | 3755 | |||
XM_017004570.1 | XP_016860059.1 | 22, 23 | 3755 | |||
XM_017004571.1 | XP_016860060.1 | 22, 23 | 3693 | |||
XM_017004572.1 | XP_016860061.1 | 20, 21 | 3343 | |||
GenBank mRNA | AB046783.2 | - | 22, 23 | 3772 | ||
AB053305.1 | - | 23, 24 | 3818 | |||
AF391100.1 | - | 23, 24 | 3818 |