Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020919.3 | NP_065970.2 | 29 | 4864 | ||
XM_006712654.2 | XP_006712717.1 | 29 | 5441 | |||
XM_006712655.3 | XP_006712718.1 | 25 | 3078 | |||
XM_011511530.2 | XP_011509832.1 | 28 | 4576 | |||
XM_017004569.1 | XP_016860058.1 | 28 | 4565 | |||
XM_017004571.1 | XP_016860060.1 | 28 | 4503 | |||
XM_017004572.1 | XP_016860061.1 | 26 | 4156 | |||
GenBank mRNA | AB046783.2 | - | 28 | 4585 | ||
AB053305.1 | - | 29 | 4631 | |||
AF391100.1 | - | 29 | 4631 | |||
AK056413.1 | - | 21 | 2612 |