Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETD1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015048.1 | NP_055863.1 | 2 | 205 | ||
XM_005253858.4 | XP_005253915.1 | 3 | 364 | |||
XM_006719296.3 | XP_006719359.1 | 3 | 280 | |||
GenBank mRNA | JF813787.1 | - | 2 | 199 |