Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136115.2 | NP_001129587.1 | 3 | 525 | ||
NM_015176.3 | NP_055991.1 | 3 | 525 | |||
GenBank mRNA | AB007952.1 | - | 3 | 276 | ||
AK001628.1 | - | 3 | 501 | |||
AK307892.1 | - | 3 | 497 | |||
BC031576.1 | - | 3 | 494 | 5188219 | ||
DB136175.1 | - | 3 | 526 |