Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYCE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105578.1 | NP_001099048.1 | 3 | 220 | ||
XM_005259848.4 | XP_005259905.1 | 3 | 292 | |||
XM_011527882.2 | XP_011526184.1 | 3 | 622 | |||
XM_011527883.2 | XP_011526185.1 | 3 | 622 | |||
GenBank mRNA | AK097443.1 | - | 2 | 1473 | ||
AK302373.1 | - | 3 | 210 | |||
BC035819.2 | - | 2 | 189 | |||
DB449708.1 | - | 3 | 224 |