Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145808.3 | NP_665807.1 | 4 | 3292 | ||
GenBank mRNA | AK055660.1 | - | 1 | 1843 | ||
AL110137.1 | - | 1 | 671 | |||
AL834231.1 | - | 4 | 3164 | |||
AY611627.1 | - | 1 | 490 |
Gene Symbol: LUZP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128619.2 | NP_001122091.2 | 4 | 3292 |