Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDFI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300804.1 | NP_001287733.1 | 4, 5 | 597 | ||
NM_001300806.1 | NP_001287735.1 | 2, 3 | 546 | |||
NM_005586.3 | NP_005577.1 | 3, 4 | 460 | |||
XM_005249117.3 | XP_005249174.1 | 3, 4 | 463 | |||
XM_011514625.2 | XP_011512927.1 | 4, 5 | 748 | |||
XM_017010867.1 | XP_016866356.1 | 2, 3 | 1119 | |||
GenBank mRNA | AK290735.1 | - | 2, 3 | 544 | ||
BC007836.2 | - | 3, 4 | 409 | 4110841 | ||
BG818571.1 | - | 3, 4 | 459 | |||
BJ989485.1 | - | 2, 3 | 537 | |||
BX397111.2 | - | 4, 5 | 580 | |||
CA393148.1 | - | 3, 4 | 451 | |||
HY007825.1 | - | 482 |