Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUMF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042469.1 | NP_001035934.2 | 2 | 263 | ||
NM_001042470.1 | NP_001035935.2 | 2 | 263 | |||
NM_001130069.2 | NP_001123541.1 | 2 | 263 | |||
NM_015411.2 | NP_056226.2 | 2 | 263 | |||
XM_011515254.2 | XP_011513556.1 | 2 | 263 | |||
XM_017011939.1 | XP_016867428.1 | 2 | 263 | |||
XM_017011940.1 | XP_016867429.1 | 2 | 480 | |||
GenBank mRNA | AK075477.1 | - | 2 | 222 | ||
AK297793.1 | - | 2 | 177 | |||
AK298605.1 | - | 2 | 203 | |||
AK301627.1 | - | 2 | 180 | |||
AL050037.1 | - | 2 | 179 | |||
AY323911.1 | - | 2 | 175 | |||
AY359103.1 | - | 2 | 193 | |||
BC006159.1 | - | 2 | 161 | |||
BC015600.2 | - | 2 | 160 | |||
BC065222.1 | - | 2 | 210 | |||
BC084539.1 | - | 2 | 193 | 6599080 | ||
BC111092.1 | - | 2 | 216 | 40068651 | ||
BM423367.1 | - | 2 | 165 | |||
BP296294.1 | - | 2 | 485 | |||
CF131206.1 | - | 2 | 215 | |||
CR936749.1 | - | 2 | 198 | |||
CR936757.1 | - | 2 | 206 | |||
CX164099.1 | - | 2 | 141 | |||
DA130450.1 | - | 2 | 232 | |||
DA924174.1 | - | 2 | 264 | |||
DB010869.1 | - | 2 | 181 | |||
KU178582.1 | - | 1 | 88 | |||
KU178583.1 | - | 1 | 88 | |||
KU178584.1 | - | 1 | 88 | |||
KU178585.1 | - | 1 | 88 |