Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UHRF1BP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006947.1 | NP_001006948.1 | 4, 5 | 568 | ||
NM_015054.1 | NP_055869.1 | 4, 5 | 568 | |||
XM_005268738.2 | XP_005268795.1 | 5, 6 | 674 | |||
XM_005268739.3 | XP_005268796.1 | 4, 5 | 528 | |||
XM_017019047.1 | XP_016874536.1 | 4, 5 | 528 | |||
XM_017019048.1 | XP_016874537.1 | 5, 6 | 674 | |||
XM_017019049.1 | XP_016874538.1 | 4, 5 | 528 | |||
XM_017019050.1 | XP_016874539.1 | 4, 5 | 528 | |||
XM_017019051.1 | XP_016874540.1 | 5, 6 | 674 | |||
GenBank mRNA | AK125813.1 | - | 4, 5 | 528 | ||
BC014891.1 | - | 4, 5 | 551 | |||
BC127017.1 | - | 4, 5 | 386 | 40124660 |