Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114380.1 | NP_001107852.1 | 21 | 2566 | ||
NM_002209.2 | NP_002200.2 | 23 | 2818 | |||
XM_005255313.1 | XP_005255370.1 | 23 | 2735 | |||
XM_006721044.1 | XP_006721107.1 | 21 | 2520 | |||
XM_011545849.1 | XP_011544151.1 | 23 | 2751 | |||
GenBank mRNA | AK303190.1 | - | 4, 5 | 431 | ||
AK314246.1 | - | 23 | 2772 | |||
BC008777.2 | - | 21 | 2499 | 3142951 | ||
Y00796.1 | - | 23 | 2730 |