Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SWAP70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297714.1 | NP_001284643.1 | 3 | 531 | ||
NM_015055.3 | NP_055870.2 | 4 | 705 | |||
XM_005252830.4 | XP_005252887.1 | 2 | 606 | |||
GenBank mRNA | AB014540.1 | - | 4 | 657 | ||
AF134894.1 | - | 4 | 676 | |||
AF210818.1 | - | 4 | 602 | |||
AK096859.1 | - | 3 | 531 | |||
AK304104.1 | - | 4 | 569 | |||
BC000616.2 | - | 4 | 678 | 3346240 |