Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3R3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114172.1 | NP_001107644.1 | 2 | 326 | ||
NM_001303428.1 | NP_001290357.1 | 2 | 170 | |||
NM_001303429.1 | NP_001290358.1 | 1 | 825 | |||
NM_003629.3 | NP_003620.3 | 1 | 825 | |||
XM_017002614.1 | XP_016858103.1 | 2 | 203 | |||
GenBank mRNA | AK313726.1 | - | 2 | 338 | ||
BC005381.1 | - | 1 | 296 | |||
BC021622.1 | - | 1 | 759 | 3910221 | ||
BX537388.1 | - | 2 | 170 | |||
CR749649.1 | - | 1 | 746 | |||
D88532.1 | - | 1 | 489 |