Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134693.1 | NP_001128165.1 | 1, 2 | 107 | ||
GenBank mRNA | AL049447.1 | - | 1, 2 | 33 | ||
BC105283.1 | - | 1, 2 | 65 | |||
BF697310.1 | - | 1, 2 | 107 |