Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018989.1 | NP_061862.1 | 8 | 1333 | ||
XM_005268466.1 | XP_005268523.1 | 8 | 1386 | |||
XM_005268467.1 | XP_005268524.1 | 8 | 1386 | |||
XM_005268468.1 | XP_005268525.1 | 8 | 1383 | |||
XM_006714787.1 | XP_006714850.1 | 9 | 1350 | |||
XM_011537661.1 | XP_011535963.1 | 8 | 1386 | |||
XM_011537662.1 | XP_011535964.1 | 8 | 1383 | |||
XM_017009587.1 | XP_016865076.1 | 9 | 1347 | |||
GenBank mRNA | AB037732.1 | - | 4 | 656 | ||
AK128738.1 | - | 8 | 1383 | |||
BX647384.1 | - | 8 | 1333 |