Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031725.5 | NP_001026895.2 | 8 | 2039 | ||
NM_001320182.1 | NP_001307111.1 | 5 | 1465 | |||
XM_005245519.1 | XP_005245576.1 | 7 | 1819 | |||
XM_005245521.2 | XP_005245578.1 | 7 | 1696 | |||
XM_011510035.2 | XP_011508337.1 | 8 | 1972 | |||
XM_017002431.1 | XP_016857920.1 | 8 | 2727 | |||
XM_017002432.1 | XP_016857921.1 | 8 | 2212 | |||
GenBank mRNA | BC041801.1 | - | 8 | 2030 | 5302098 |