Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF707 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100598.1 | NP_001094068.1 | 6 | 528 | ||
NM_001100599.1 | NP_001094069.1 | 5 | 430 | |||
NM_001288805.1 | NP_001275734.1 | 8 | 1242 | |||
NM_001288806.1 | NP_001275735.1 | 6 | 528 | |||
NM_001288807.1 | NP_001275736.1 | 6 | 516 | |||
NM_001288808.1 | NP_001275737.1 | 4 | 363 | |||
NM_001288809.1 | NP_001275738.1 | 4 | 537 | |||
NM_173831.3 | NP_776192.2 | 7 | 602 | |||
NR_110190.1 | 5 | 463 | ||||
NR_110191.1 | 6 | 561 | ||||
NR_110192.1 | 5 | 466 | ||||
NR_110193.1 | 6 | 561 | ||||
XM_005250883.3 | XP_005250940.1 | 6 | 549 | |||
XM_006716542.3 | XP_006716605.1 | 7 | 618 | |||
XM_006716543.3 | XP_006716606.1 | 6 | 523 | |||
XM_011516977.2 | XP_011515279.1 | 6 | 707 | |||
XM_011516979.2 | XP_011515281.1 | 7 | 568 | |||
XM_017013335.1 | XP_016868824.1 | 8 | 725 | |||
GenBank mRNA | AK001126.1 | - | 5 | 459 | ||
AK055198.1 | - | 3 | 1348 | |||
AK055263.1 | - | 5 | 504 | |||
AK290432.1 | - | 6 | 499 | |||
DA007942.1 | - | 6 | 499 |