Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002817.3 | NP_002808.3 | 7 | 727 | ||
NM_175932.2 | NP_787128.2 | 5 | 733 | |||
XM_011520235.2 | XP_011518537.1 | 7 | 860 | |||
GenBank mRNA | AB009398.1 | - | 7 | 554 | ||
AF083245.1 | - | 7 | 539 | |||
AF086708.1 | - | 7 | 485 | |||
AF107837.1 | - | 7 | 518 | |||
AK094775.1 | - | 5 | 564 | |||
AK222895.1 | - | 7 | 558 | |||
AK295944.1 | - | 6 | 524 | |||
AK307447.1 | - | 6 | 452 | |||
AK314733.1 | - | 7 | 989 | |||
BC001100.2 | - | 7 | 540 | 3506530 | ||
BC001747.1 | - | 7 | 508 | 3535807 | ||
BM456586.1 | - | 7 | 558 | |||
BT007307.1 | - | 7 | 485 | |||
BX647314.1 | - | 6 | 462 |