Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002817.3 | NP_002808.3 | 6 | 600 | ||
NM_175932.2 | NP_787128.2 | 4 | 606 | |||
XM_011520235.2 | XP_011518537.1 | 6 | 733 | |||
GenBank mRNA | AB009398.1 | - | 6 | 427 | ||
AF083245.1 | - | 6 | 412 | |||
AF086708.1 | - | 6 | 358 | |||
AF107837.1 | - | 6 | 391 | |||
AK094775.1 | - | 4 | 437 | |||
AK222895.1 | - | 6 | 431 | |||
AK295944.1 | - | 5 | 397 | |||
AK307447.1 | - | 5 | 325 | |||
AK314733.1 | - | 6 | 862 | |||
BC001100.2 | - | 6 | 413 | 3506530 | ||
BC001747.1 | - | 6 | 381 | 3535807 | ||
BM456586.1 | - | 6 | 431 | |||
BT007307.1 | - | 6 | 358 | |||
BX647314.1 | - | 5 | 335 |