Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098834.2 | NP_001092304.1 | 2 | 705 | ||
XM_017011964.1 | XP_016867453.1 | 3 | 722 | |||
XM_017011965.1 | XP_016867454.1 | 3 | 719 |