Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SVIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320340.1 | NP_001307269.1 | 4 | 769 | ||
NM_001320342.1 | NP_001307271.1 | 4 | 653 | |||
NM_148893.2 | NP_683691.1 | 4 | 660 | |||
NR_135213.1 | 4 | 615 | ||||
GenBank mRNA | AF527534.1 | - | 4 | 634 | ||
AL833119.1 | - | 4 | 633 | |||
BG531443.1 | - | 4 | 598 |