Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112704.1 | NP_001106175.1 | 1 | 318 | ||
NM_199131.2 | NP_954582.1 | 1 | 318 | |||
GenBank mRNA | AK127095.1 | - | 1 | 315 | ||
BC101694.1 | - | 1 | 102 | 8069200 | ||
BC101696.1 | - | 1 | 102 | 8069202 |