Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007138.1 | NP_009069.1 | 4 | 1037 | ||
GenBank mRNA | BC137211.1 | - | 4 | 964 | 9021208 |