Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021014.3 | NP_066294.1 | 6 | 504 | ||
XM_011543885.2 | XP_011542187.1 | 6 | 457 | |||
GenBank mRNA | AK310513.1 | - | 6 | 457 | ||
BC005904.1 | - | 6 | 458 | 4064072 | ||
BC103862.1 | - | 6 | 503 | 40003336 | ||
S82471.1 | - | 5 | 364 | |||
U90840.1 | - | 6 | 417 |