Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDFY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020945.1 | NP_065996.1 | 60 | 9381 | ||
XM_005270004.3 | XP_005270061.1 | 60 | 9496 | |||
XM_011539986.2 | XP_011538288.2 | 60 | 9499 | |||
XM_011539987.2 | XP_011538289.2 | 60 | 9481 | |||
XM_011539988.2 | XP_011538290.1 | 61 | 9580 | |||
XM_017016463.1 | XP_016871952.1 | 61 | 9423 | |||
XM_017016464.1 | XP_016871953.1 | 42 | 5810 | |||
GenBank mRNA | AB046827.1 | - | 28 | 3564 | ||
AK024502.1 | - | 10 | 3938 | |||
AK074085.1 | - | 41 | 5439 | |||
AK074443.1 | - | 23 | 2784 | |||
AK301736.1 | - | 14 | 1837 | |||
AL833669.1 | - | 21 | 2510 | |||
BC015694.2 | - | 5 | 712 | |||
BC032420.1 | - | 5 | 800 | |||
BC047574.1 | - | 21 | 2450 |