Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDFY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020945.1 | NP_065996.1 | 43 | 7215 | ||
XM_005270004.3 | XP_005270061.1 | 43 | 7330 | |||
XM_011539986.2 | XP_011538288.2 | 43 | 7333 | |||
XM_011539987.2 | XP_011538289.2 | 43 | 7336 | |||
XM_011539988.2 | XP_011538290.1 | 44 | 7414 | |||
XM_011539990.2 | XP_011538292.1 | 43 | 7361 | |||
XM_017016463.1 | XP_016871952.1 | 44 | 7257 | |||
XM_017016464.1 | XP_016871953.1 | 25 | 3644 | |||
GenBank mRNA | AB046827.1 | - | 10 | 1398 | ||
AK074085.1 | - | 23 | 3273 | |||
AK074443.1 | - | 5 | 618 | |||
AL833669.1 | - | 3 | 344 | |||
BC047574.1 | - | 3 | 284 |