Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDFY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020945.1 | NP_065996.1 | 50 | 7926 | ||
XM_005270004.3 | XP_005270061.1 | 50 | 8041 | |||
XM_011539986.2 | XP_011538288.2 | 50 | 8044 | |||
XM_011539987.2 | XP_011538289.2 | 50 | 8047 | |||
XM_011539988.2 | XP_011538290.1 | 51 | 8125 | |||
XM_017016463.1 | XP_016871952.1 | 51 | 7968 | |||
XM_017016464.1 | XP_016871953.1 | 32 | 4355 | |||
GenBank mRNA | AB046827.1 | - | 18 | 2109 | ||
AK024502.1 | - | 1 | 93 | |||
AK074085.1 | - | 31 | 3984 | |||
AK074443.1 | - | 13 | 1329 | |||
AK301736.1 | - | 4 | 382 | |||
AK310374.1 | - | 4 | 426 | |||
AL833669.1 | - | 11 | 1055 | |||
BC047574.1 | - | 11 | 995 |