Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AVL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015060.2 | NP_055875.1 | 10 | 1115 | ||
XM_005249668.2 | XP_005249725.1 | 10 | 1052 | |||
XM_005249669.2 | XP_005249726.1 | 10 | 1052 | |||
XM_005249671.4 | XP_005249728.1 | 10 | 1052 | |||
XM_011515227.2 | XP_011513529.1 | 10 | 1052 | |||
GenBank mRNA | AK090619.1 | - | 10 | 977 | ||
BC027724.2 | - | 10 | 967 | 4811237 | ||
D87682.1 | - | 8 | 624 |