Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM193B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190946.2 | NP_001177875.1 | 6 | 1880 | ||
NR_024019.3 | 9 | 3539 | ||||
XM_006714875.2 | XP_006714938.1 | 7 | 2008 | |||
XM_006714876.1 | XP_006714939.1 | 7 | 2731 | |||
XM_006714878.1 | XP_006714941.1 | 8 | 2585 | |||
XM_006714879.3 | XP_006714942.1 | 8 | 1828 | |||
XM_006714880.1 | XP_006714943.1 | 7 | 2626 | |||
XM_006714884.1 | XP_006714947.1 | 5 | 1129 | |||
XM_006714885.1 | XP_006714948.1 | 4 | 933 | |||
XM_011534572.1 | XP_011532874.1 | 7 | 1903 | |||
XM_011534573.1 | XP_011532875.1 | 8 | 2003 | |||
XM_011534574.1 | XP_011532876.1 | 8 | 2448 | |||
XM_011534579.2 | XP_011532881.1 | 7 | 1767 | |||
XM_011534580.1 | XP_011532882.1 | 8 | 2792 | |||
XM_011534581.1 | XP_011532883.1 | 7 | 2472 | |||
XM_011534582.1 | XP_011532884.1 | 8 | 2979 | |||
XM_011534583.1 | XP_011532885.1 | 8 | 2720 | |||
GenBank mRNA | AB067518.1 | - | 9 | 3496 | ||
AK001266.1 | - | 3 | 809 | |||
AK096032.1 | - | 7 | 1691 | |||
AL833096.1 | - | 1 | 760 | |||
BC008784.2 | - | 5 | 1129 | 3626782 | ||
BC071696.1 | - | 3 | 783 | |||
BC086859.1 | - | 5 | 1353 | |||
BX647587.1 | - | 6 | 1624 |