Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015559.2 | NP_056374.2 | 4 | 3433 | ||
GenBank mRNA | AB007897.2 | - | 4 | 3433 | ||
AB022660.1 | - | 3 | 2980 | |||
BC146776.1 | - | 4 | 3433 | 9007225 |