Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGCX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000821.6 | NP_000812.2 | 13, 14 | 1986 | ||
NM_001142269.3 | NP_001135741.1 | 12, 13 | 1815 | |||
XM_011532764.2 | XP_011531066.1 | 12, 13 | 1781 | |||
XM_011532765.2 | XP_011531067.1 | 11, 12 | 1620 | |||
GenBank mRNA | AK130139.1 | - | 4, 5 | 578 | ||
AK222827.1 | - | 13, 14 | 1946 | |||
AK225663.1 | - | 13, 14 | 1945 | |||
AK297397.1 | - | 12, 13 | 1755 | |||
AK313126.1 | - | 13, 14 | 1954 | |||
BC013979.2 | - | 13, 14 | 1908 | 4129593 | ||
EU847509.1 | - | 13, 14 | 1883 | |||
L17128.1 | - | 13, 14 | 2035 | |||
M81592.1 | - | 13, 14 | 1966 |