Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015131.2 | NP_055946.1 | 5 | 742 | ||
GenBank mRNA | AK000836.1 | - | 4 | 438 | ||
BC033206.1 | - | 4 | 447 | |||
D26488.1 | - | 5 | 715 | |||
D87716.1 | - | 3 | 221 |