Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF5AL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099692.1 | NP_001093162.1 | 1 | 154 | ||
GenBank mRNA | BC070048.1 | - | 1 | 154 |