Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BICDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103175.1 | NP_001096645.1 | 3 | 559 | ||
XM_005255135.4 | XP_005255192.1 | 4 | 620 | |||
XM_011522391.1 | XP_011520693.1 | 3 | 808 | |||
GenBank mRNA | AK307181.1 | - | 2 | 103 | ||
AL833717.1 | - | 2 | 1056 | |||
AL833749.1 | - | 2 | 2084 | |||
BC128602.1 | - | 3 | 559 | 40132962 |