Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BICDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103175.1 | NP_001096645.1 | 1 | 317 | ||
XM_005255135.4 | XP_005255192.1 | 2 | 378 | |||
GenBank mRNA | BC128602.1 | - | 1 | 317 | 40132962 | |
BG142147.1 | - | 1 | 278 |