Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122674.1 | NP_001116146.1 | 6 | 574 | ||
NM_002858.3 | NP_002849.1 | 6 | 574 | |||
XM_006710802.2 | XP_006710865.2 | 7 | 937 | |||
XM_017001952.1 | XP_016857441.1 | 6 | 1115 | |||
XM_017001953.1 | XP_016857442.1 | 6 | 495 | |||
XM_017001954.1 | XP_016857443.1 | 5 | 555 | |||
GenBank mRNA | AK296791.1 | - | 7 | 610 | ||
AK302714.1 | - | 5 | 499 | |||
AK308159.1 | - | 6 | 535 | |||
BC009712.2 | - | 6 | 512 | |||
BC068509.1 | - | 6 | 535 | 5265201 | ||
BT006644.1 | - | 6 | 472 | |||
M81182.1 | - | 6 | 472 | |||
X58528.1 | - | 6 | 496 |