Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ERC1 SIRNA Assays ›
Gene Symbol:
ERC1
Gene Name:
ELKS/RAB6-interacting/CAST family member 1
Gene Aliases:
Cast2, ELKS, ERC-1, RAB6IP2
Chromosome Location:
Chr.12: 990010 - 1495933 on Build GRCh38
UniGene ID:
Hs.601216
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s22995
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ERC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001301248.1
NP_001288177.1
9
2099
NM_178039.3
NP_829883.1
9
2130
NM_178040.3
NP_829884.1
10
2214
NR_027946.2
9
2295
NR_027948.2
9
2295
NR_027949.2
5
1314
XM_011520941.2
XP_011519243.1
10
2258
XM_017019052.1
XP_016874541.1
10
2333
XM_017019053.1
XP_016874542.1
10
2267
XM_017019054.1
XP_016874543.1
10
2193
XM_017019055.1
XP_016874544.1
10
2189
XM_017019056.1
XP_016874545.1
10
2130
XM_017019057.1
XP_016874546.1
10
2324
XM_017019058.1
XP_016874547.1
10
2258
XM_017019059.1
XP_016874548.1
10
2333
XM_017019060.1
XP_016874549.1
10
2324
XM_017019061.1
XP_016874550.1
10
2333
XM_017019062.1
XP_016874551.1
10
2324
XM_017019063.1
XP_016874552.1
10
2333
XM_017019064.1
XP_016874553.1
10
2324
XM_017019065.1
XP_016874554.1
9
2240
XM_017019066.1
XP_016874555.1
9
2240
XM_017019067.1
XP_016874556.1
9
2240
XM_017019068.1
XP_016874557.1
10
2324
XM_017019069.1
XP_016874558.1
10
2333
XM_017019070.1
XP_016874559.1
10
2324
XM_017019071.1
XP_016874560.1
9
2240
XM_017019072.1
XP_016874561.1
10
2333
XM_017019073.1
XP_016874562.1
10
2333
XM_017019074.1
XP_016874563.1
3
313
XM_017019075.1
XP_016874564.1
3
311
GenBank mRNA
AB015617.1
-
9
2253
AB029004.2
-
8
1982
AB053468.1
-
9
2236
AB053469.1
-
5
1249
AB053470.1
-
9
2072
AB053471.1
-
10
2124
AK091577.1
-
9
2240
AK294351.1
-
7
1214
AK309992.1
-
7
874
BC068006.1
-
8
1981
BC132782.1
-
9
2075
40146804
BC132784.1
-
9
2075
40146806
BC144287.1
-
9
2075
BC150248.1
-
8
2003
9007473
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
I-kappaB phosphorylation
multicellular organism development
protein transport
retrograde transport, endosome to Golgi
positive regulation of NF-kappaB transcription factor activity
cell-cell adhesion
protein binding
Rab GTPase binding
PDZ domain binding
leucine zipper domain binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-pg4v9:80/100.66.77.47:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline