Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZEB1 SIRNA Assays ›
Gene Symbol:
ZEB1
Gene Name:
zinc finger E-box binding homeobox 1
Gene Aliases:
AREB6, BZP, DELTAEF1, FECD6, NIL2A, PPCD3, TCF8, ZFHEP, ZFHX1A
Chromosome Location:
Chr.10: 31318495 - 31529814 on Build GRCh38
UniGene ID:
Hs.124503
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s229970
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZEB1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128128.2
NP_001121600.1
9
3321
NM_001174093.1
NP_001167564.1
8
2981
NM_001174094.1
NP_001167565.1
9
3318
NM_001174095.1
NP_001167566.1
8
2840
NM_001174096.1
NP_001167567.1
9
3044
NM_001323638.1
NP_001310567.1
13
3691
NM_001323641.1
NP_001310570.1
9
3271
NM_001323642.1
NP_001310571.1
11
3573
NM_001323643.1
NP_001310572.1
11
3351
NM_001323644.1
NP_001310573.1
8
3070
NM_001323645.1
NP_001310574.1
10
3414
NM_001323646.1
NP_001310575.1
12
3652
NM_001323647.1
NP_001310576.1
10
3184
NM_001323648.1
NP_001310577.1
9
3124
NM_001323649.1
NP_001310578.1
10
3187
NM_001323650.1
NP_001310579.1
12
3419
NM_001323651.1
NP_001310580.1
10
3087
NM_001323652.1
NP_001310581.1
11
3236
NM_001323653.1
NP_001310582.1
11
3343
NM_001323654.1
NP_001310583.1
11
3230
NM_001323655.1
NP_001310584.1
11
3340
NM_001323656.1
NP_001310585.1
9
3015
NM_001323657.1
NP_001310586.1
9
3018
NM_001323658.1
NP_001310587.1
9
2857
NM_001323659.1
NP_001310588.1
9
3075
NM_001323660.1
NP_001310589.1
9
3072
NM_001323661.1
NP_001310590.1
10
3218
NM_001323662.1
NP_001310591.1
11
3620
NM_001323663.1
NP_001310592.1
12
3559
NM_001323664.1
NP_001310593.1
11
3543
NM_001323665.1
NP_001310594.1
10
3461
NM_001323666.1
NP_001310595.1
9
3401
NM_001323671.1
NP_001310600.1
10
3297
NM_001323672.1
NP_001310601.1
11
3349
NM_001323673.1
NP_001310602.1
10
3294
NM_001323674.1
NP_001310603.1
7
2819
NM_001323675.1
NP_001310604.1
7
2929
NM_001323676.1
NP_001310605.1
9
3154
NM_001323677.1
NP_001310606.1
9
3151
NM_001323678.1
NP_001310607.1
7
3096
NM_030751.5
NP_110378.3
9
3041
XM_006717498.2
XP_006717561.1
12
3479
XM_011519643.2
XP_011517945.1
12
3782
XM_017016597.1
XP_016872086.1
12
3381
XM_017016598.1
XP_016872087.1
12
6511
XM_017016599.1
XP_016872088.1
11
6349
XM_017016600.1
XP_016872089.1
13
6590
XM_017016601.1
XP_016872090.1
12
6508
XM_017016602.1
XP_016872091.1
13
6587
XM_017016603.1
XP_016872092.1
12
6425
GenBank mRNA
AK091478.1
-
8
2941
AK294775.1
-
9
3236
AK300830.1
-
8
2788
AK314683.1
-
9
3002
AL831979.1
-
11
3299
BC009572.1
-
9
2997
BC112392.1
-
9
2997
40036600
BX647794.1
-
13
3692
M81699.1
-
3
1913
U12170.1
-
9
3002
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
immune response
pattern specification process
central nervous system development
cell proliferation
negative regulation of cell proliferation
regulation of mesenchymal cell proliferation
regulation of transforming growth factor beta receptor signaling pathway
negative regulation of epithelial cell differentiation
regulation of T cell differentiation in thymus
positive regulation of neuron differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
embryonic camera-type eye morphogenesis
embryonic skeletal system morphogenesis
semicircular canal morphogenesis
regulation of smooth muscle cell differentiation
cartilage development
cellular response to amino acid stimulus
cochlea morphogenesis
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
transcription corepressor activity
protein binding
transcription factor binding
zinc ion binding
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-77psw:80/100.66.77.247:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline