Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173595.3 | NP_775866.2 | 6, 7 | 649 | ||
XM_011538197.2 | XP_011536499.1 | 5, 6 | 751 | |||
XM_017019183.1 | XP_016874672.1 | 5, 6 | 832 | |||
XM_017019184.1 | XP_016874673.1 | 6, 7 | 681 | |||
GenBank mRNA | AB190990.1 | - | 6, 7 | 666 | ||
AK125379.1 | - | 6, 7 | 666 |